Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000300900.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000585705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000585705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000585705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000585705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000585705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000587265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000587265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000590203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
ENST00000591725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012289.hg.1 |
NM_000717.4 |
Refseq |