Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055236.hg.1 |
TC01003565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055236.hg.1 |
ENST00000234816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055236.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055236.hg.1 |
ENST00000415564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055236.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055238.hg.1 |
TC01003565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055238.hg.1 |
ENST00000234816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055238.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055238.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055235.hg.1 |
TC01003565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055235.hg.1 |
ENST00000234816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055235.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055235.hg.1 |
ENST00000415564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055235.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055239.hg.1 |
TC01003565.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055239.hg.1 |
ENST00000234816.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055239.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055239.hg.1 |
ENST00000415564.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055239.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225689.hg.1 |
TC0100016539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225691.hg.1 |
TC0100016539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100225690.hg.1 |
TC0100016539.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000234816.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000367629.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
ENST00000444255.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016539.hg.1 |
NM_004673.3 |
Refseq |