Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009337.hg.1 |
TC08000689.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009337.hg.1 |
ENST00000297347.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009337.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000519879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
ENST00000522839.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_001282986.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008641.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
ENST00000297347.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
ENST00000519391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
NM_080651.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159285.hg.1 |
TC0800008641.hg.1 |
Clariom D TC |