Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000301659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000577447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000577447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000577447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
ENST00000577447.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007771.hg.1 |
NM_178171.4 |
Refseq |