Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080463.1 | NP_001073932.1 | 36 - 37 | 5818 | 107 | |
NM_001377.2 | NP_001368.2 | 36 - 37 | 5818 | 107 | ||
XM_006718903.2 | XP_006718966.1 | 36 - 37 | 5818 | 107 | ||
XM_017018291.1 | XP_016873780.1 | 36 - 37 | 5818 | 107 | ||
XM_017018292.1 | XP_016873781.1 | 33 - 34 | 5072 | 107 | ||
XM_017018293.1 | XP_016873782.1 | 36 - 37 | 5818 | 107 | ||
GenBank mRNA | AB290167.1 | - | 36 - 37 | 5872 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |