Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014976.1 | NP_055791.1 | 34 - 35 | 5367 | 62 | |
XM_005269647.3 | XP_005269704.1 | 34 - 35 | 5410 | 62 | ||
XM_011539538.2 | XP_011537840.1 | 34 - 35 | 5358 | 62 | ||
XM_011539539.2 | XP_011537841.1 | 34 - 35 | 5355 | 62 | ||
XM_011539540.1 | XP_011537842.1 | 33 - 34 | 5224 | 62 | ||
GenBank mRNA | D80007.1 | - | 34 - 35 | 5320 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |