Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021190.hg.1 |
ENST00000227524.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021190.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021190.hg.1 |
TC11001844.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021191.hg.1 |
ENST00000227524.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021191.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11021191.hg.1 |
TC11001844.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183065.hg.1 |
TC1100011010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183067.hg.1 |
TC1100011010.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000540473.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000541371.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
ENST00000546152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011010.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000227524.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000535326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
ENST00000539960.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
NM_014502.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100183066.hg.1 |
TC1100011010.hg.1 |
Clariom D TC |