Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
TC07000650.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000292535.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000360264.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000546411.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000549414.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000550008.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
ENST00000556210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
NM_001202543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07010152.hg.1 |
NM_181552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000292535.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000360264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000546411.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000549414.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000550008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000556210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
NM_001202543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
NM_181552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000292535.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000360264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000546411.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000549414.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000550008.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
ENST00000556210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
NM_001202543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
NM_181552.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700160865.hg.1 |
TC0700008597.hg.1 |
Clariom D TC |