Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024763.4 | NP_079039.4 | 14 - 15 | 2169 | 143 | |
XM_005271212.3 | XP_005271269.1 | 13 - 14 | 1927 | 143 | ||
XM_011542161.2 | XP_011540463.1 | 13 - 14 | 2035 | 143 | ||
XM_017002355.1 | XP_016857844.1 | 11 - 12 | 1542 | 143 | ||
XM_017002356.1 | XP_016857845.1 | 12 - 13 | 1786 | 143 | ||
GenBank mRNA | AK127011.1 | - | 11 - 12 | 2538 | 143 | |
AK292830.1 | - | 15 - 16 | 2122 | 143 | ||
AL133617.1 | - | 5 - 6 | 565 | 143 | ||
BC126379.1 | - | 14 - 15 | 2144 | 8992095 | 143 | |
BC126381.1 | - | 14 - 15 | 2144 | 8992097 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |