Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001402.hg.1 |
TC07000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001402.hg.1 |
ENST00000405863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001402.hg.1 |
ENST00000417186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001402.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001401.hg.1 |
TC07000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001401.hg.1 |
ENST00000405863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001401.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001400.hg.1 |
TC07000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001400.hg.1 |
ENST00000405863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001400.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001405.hg.1 |
TC07000083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001405.hg.1 |
ENST00000405863.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001405.hg.1 |
ENST00000417186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07001405.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000497400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
ENST00000602349.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006690.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146291.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146291.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146291.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146291.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146291.hg.1 |
TC0700006690.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146290.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146290.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146290.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146290.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146290.hg.1 |
TC0700006690.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000405863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000429542.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
ENST00000438764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
NM_152745.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700146294.hg.1 |
TC0700006690.hg.1 |
Clariom D TC |