Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008103.hg.1 |
TC06000677.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008103.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008103.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008323.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
ENST00000370862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
ENST00000615358.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
NM_001526.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600158107.hg.1 |
TC0600008323.hg.1 |
Clariom D TC |