Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171851.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171851.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171851.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171851.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171851.hg.1 |
TC0300009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300171852.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171852.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171852.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300171852.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300171852.hg.1 |
TC0300009880.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000414920.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
ENST00000530635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009880.hg.1 |
NR_046596.1 |
Refseq |