Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592589.2 | XP_005592646.1 | 1 - 2 | 838 | 96 | |
XM_015442958.1 | XP_015298444.1 | 1 - 2 | 838 | 96 | ||
GenBank mRNA | AB168345.1 | - | 1 - 2 | 777 | 96 | |
AB169914.1 | - | 1 - 2 | 808 | 96 | ||
AB170847.1 | - | 1 - 2 | 778 | 96 | ||
AB171109.1 | - | 779 | 96 | |||
AB171183.1 | - | 1 - 2 | 808 | 96 | ||
AB171260.1 | - | 1 - 2 | 793 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |