Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287700.1 | NP_001274629.1 | 1 - 2 | 65 | 76 | |
XM_005580128.2 | XP_005580185.1 | 3 - 4 | 620 | 76 | ||
XM_005580130.2 | XP_005580187.1 | 2 - 3 | 284 | 76 | ||
GenBank mRNA | AB446482.1 | - | 1 - 2 | 65 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Inventoried Probe spans exons |