Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tktl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031379.2 | NP_113556.2 | 8 - 9 | 1210 | 85 | |
GenBank mRNA | AF285571.1 | - | 8 - 9 | 1185 | 85 | |
AJ627042.1 | - | 1 - 2 | 100 | 85 | ||
AK029546.1 | - | 8 - 9 | 1211 | 85 | ||
AK139841.1 | - | 8 - 9 | 1210 | 85 | ||
BC113768.1 | - | 8 - 9 | 1185 | 40058456 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |