Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177397.3 | NP_796371.1 | 1 - 2 | 109 | 77 | |
GenBank mRNA | AB088359.1 | - | 1 - 2 | 109 | 77 | |
AK032887.1 | - | 1 - 2 | 109 | 77 | ||
BC107267.1 | - | 1 - 2 | 108 | 40052586 | 77 | |
BC107268.1 | - | 1 - 2 | 108 | 40052587 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |