Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126330.1 | NP_001119802.1 | 9 - 10 | 1447 | 55 | |
NM_011642.3 | NP_035772.2 | 11 - 12 | 1737 | 55 | ||
GenBank mRNA | AB221594.1 | - | 10 - 11 | 1329 | 55 | |
AK014503.1 | - | 4 - 5 | 426 | 55 | ||
AK017412.1 | - | 8 - 9 | 978 | 55 | ||
BC066045.1 | - | 9 - 10 | 1334 | 6812399 | 55 | |
BC069182.1 | - | 5 - 6 | 594 | 55 | ||
Y19234.1 | - | 11 - 12 | 1738 | 55 | ||
Y19235.1 | - | 9 - 10 | 1441 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |