Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162475.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162476.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162476.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162479.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_001162479.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_009291.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
NM_009291.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005341.mm.1 |
TC0900000725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005334.mm.1 |
NM_001162476.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005334.mm.1 |
NM_001162476.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005334.mm.1 |
TC0900000725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005336.mm.1 |
NM_001162479.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005336.mm.1 |
NM_001162479.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005336.mm.1 |
TC0900000725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005327.mm.1 |
NM_001162475.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005327.mm.1 |
NM_001162475.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005327.mm.1 |
TC0900000725.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005333.mm.1 |
NM_001162476.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005333.mm.1 |
NM_001162476.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005333.mm.1 |
TC0900000725.mm.1 |
MTA TC |