Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029734.1 | NP_084010.1 | 1 - 2 | 154 | 71 | |
GenBank mRNA | AK010831.1 | - | 1 - 2 | 175 | 71 | |
AK150578.1 | - | 1 - 2 | 206 | 71 | ||
AK167510.1 | - | 1 - 2 | 174 | 71 | ||
BC051524.1 | - | 1 - 2 | 154 | 1514917 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |