Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400018267.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018267.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018267.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018270.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018270.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018270.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018273.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018273.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018273.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177855.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177855.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018279.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018269.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018269.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018269.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177855.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177855.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018280.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018268.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018268.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018268.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177855.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177855.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018278.mm.1 |
TC0400002189.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177849.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177849.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177850.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177850.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177851.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177851.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177852.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177852.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177853.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177853.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177854.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177854.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177855.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177855.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177856.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_001177856.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_023066.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_023066.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_133723.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
NM_133723.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400018275.mm.1 |
TC0400002189.mm.1 |
MTA TC |