Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144541.1 | NP_653124.1 | 8 - 9 | 832 | 107 | |
NM_181279.1 | NP_851796.1 | 10 - 11 | 943 | 107 | ||
NM_181280.1 | NP_851797.1 | 9 - 10 | 841 | 107 | ||
NM_181281.1 | NP_851798.1 | 7 - 8 | 676 | 107 | ||
NM_181282.1 | NP_851799.1 | 8 - 9 | 753 | 107 | ||
GenBank mRNA | AF440752.1 | - | 8 - 9 | 832 | 107 | |
AF527952.1 | - | 10 - 11 | 943 | 107 | ||
AF527953.1 | - | 8 - 9 | 753 | 107 | ||
AF527954.1 | - | 8 - 9 | 763 | 107 | ||
AF527955.1 | - | 7 - 8 | 676 | 107 | ||
AF538925.1 | - | 9 - 10 | 841 | 107 | ||
AK080991.1 | - | 8 - 9 | 868 | 107 | ||
AK156929.1 | - | 8 - 9 | 924 | 107 | ||
BC061000.1 | - | 8 - 9 | 908 | 30305062 | 107 | |
BC100565.1 | - | 4 - 5 | 387 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |