Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042779.2 | NP_001036244.1 | 13 - 14 | 1534 | 54 | |
NM_001291537.1 | NP_001278466.1 | 13 - 14 | 1798 | 54 | ||
NM_001291538.1 | NP_001278467.1 | 13 - 14 | 1792 | 54 | ||
NM_001291539.1 | NP_001278468.1 | 12 - 13 | 1475 | 54 | ||
NM_009153.3 | NP_033179.2 | 11 - 12 | 1344 | 54 | ||
NR_111986.1 | 11 - 12 | 1354 | 54 | |||
GenBank mRNA | AK014333.1 | - | 11 - 12 | 1290 | 54 | |
AK033597.1 | - | 12 - 13 | 1485 | 54 | ||
AK037034.1 | - | 13 - 14 | 1523 | 54 | ||
AK043634.1 | - | 12 - 13 | 1428 | 54 | ||
AK048364.1 | - | 12 - 13 | 1756 | 54 | ||
AK148232.1 | - | 13 - 14 | 1495 | 54 | ||
AK201474.1 | - | 2 - 3 | 184 | 54 | ||
BC090669.1 | - | 12 - 13 | 1455 | 30696359 | 54 | |
BC150758.1 | - | 10 - 11 | 1190 | 9087669 | 54 | |
CN538654.1 | - | 4 - 5 | 390 | 54 | ||
X85990.1 | - | 11 - 12 | 1341 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |