Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_001168679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_001168679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_001168680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_001168680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_146010.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
NM_146010.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009931.mm.1 |
TC1000001333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_001168679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_001168679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_001168680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_001168680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_146010.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
NM_146010.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009935.mm.1 |
TC1000001333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_001168679.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_001168679.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_001168680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_001168680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_146010.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
NM_146010.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009930.mm.1 |
TC1000001333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000009932.mm.1 |
NM_001168680.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009932.mm.1 |
NM_001168680.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009932.mm.1 |
NM_146010.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009932.mm.1 |
NM_146010.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000009932.mm.1 |
TC1000001333.mm.1 |
MTA TC |