Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100016488.mm.1 |
NM_030033.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016488.mm.1 |
NM_030033.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016488.mm.1 |
TC0100002019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016486.mm.1 |
NM_030033.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016486.mm.1 |
NM_030033.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016486.mm.1 |
TC0100002019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016490.mm.1 |
NM_030033.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016490.mm.1 |
NM_030033.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016490.mm.1 |
TC0100002019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016487.mm.1 |
NM_030033.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016487.mm.1 |
NM_030033.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016487.mm.1 |
TC0100002019.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100016489.mm.1 |
NM_030033.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016489.mm.1 |
NM_030033.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100016489.mm.1 |
TC0100002019.mm.1 |
MTA TC |