Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021287.1 | NP_067262.1 | 35 - 36 | 7336 | 84 | |
GenBank mRNA | AK147671.1 | - | 35 - 36 | 7352 | 84 | |
AK172931.1 | - | 25 - 26 | 5645 | 84 | ||
BC033305.1 | - | 1 - 2 | 84 | |||
BC048851.1 | - | 13 - 14 | 84 | |||
BC079628.1 | - | 13 - 14 | 2288 | 84 | ||
BC079860.1 | - | 35 - 36 | 7336 | 6834745 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |