Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081264.1 | NP_001074733.1 | 7 - 8 | 687 | 109 | |
GenBank mRNA | AK157903.1 | - | 7 - 8 | 649 | 109 | |
AK171887.1 | - | 7 - 8 | 640 | 109 | ||
BC050854.1 | - | 7 - 8 | 652 | 6332345 | 109 | |
BC080867.1 | - | 7 - 8 | 627 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |