Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253781.1 | NP_001240710.1 | 7 - 8 | 978 | 97 | |
NM_001253782.2 | NP_001240711.1 | 6 - 7 | 787 | 97 | ||
NM_011169.5 | NP_035299.4 | 7 - 8 | 1421 | 97 | ||
GenBank mRNA | AB643813.1 | - | 5 - 6 | 747 | 97 | |
AK050317.1 | - | 7 - 8 | 1013 | 97 | ||
AK052679.1 | - | 7 - 8 | 876 | 97 | ||
AK054381.1 | - | 7 - 8 | 1241 | 97 | ||
AK054551.1 | - | 7 - 8 | 1423 | 97 | ||
AK143335.1 | - | 7 - 8 | 1108 | 97 | ||
BC005555.1 | - | 7 - 8 | 1004 | 3586308 | 97 | |
BC006652.1 | - | 7 - 8 | 895 | 3500076 | 97 | |
D10214.1 | - | 5 - 6 | 696 | 97 | ||
L13593.1 | - | 5 - 6 | 731 | 97 | ||
L14811.1 | - | 5 - 6 | 673 | 97 | ||
M22957.1 | - | 7 - 8 | 898 | 97 | ||
M22958.1 | - | 5 - 6 | 733 | 97 | ||
M22959.1 | - | 7 - 8 | 899 | 97 | ||
X73372.1 | - | 5 - 6 | 673 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |