Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201341.1 | NP_001188270.1 | 7 - 8 | 662 | 66 | |
GenBank mRNA | BC109368.1 | - | 7 - 8 | 575 | 5249862 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |