Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015771.2 | NP_056586.2 | 4 - 5 | 2204 | 88 | |
GenBank mRNA | AK029242.1 | - | 2185 | 88 | ||
AK029870.1 | - | 4 - 5 | 2185 | 88 | ||
AK029966.1 | - | 4 - 5 | 2164 | 88 | ||
AK081190.2 | - | 5 - 6 | 89 | |||
AK161243.1 | - | 3 - 4 | 1626 | 88 | ||
AK163607.1 | - | 4 - 5 | 2187 | 88 | ||
AK164789.1 | - | 4 - 5 | 2157 | 88 | ||
BC053028.1 | - | 4 - 5 | 2158 | 6409832 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |