Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285419.1 | NP_001272348.1 | 1 - 2 | 190 | 105 | |
GenBank mRNA | AY057912.1 | - | 1 - 2 | 105 | ||
EF125681.1 | - | 1 - 2 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mus musculus 1700 Inventoried Probe spans exons |