Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960660.1 | XP_027816461.1 | 1 - 2 | 1364 | 102 | |
XM_027960661.1 | XP_027816462.1 | 2 - 3 | 2426 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |