Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843651.1 | XP_028699484.1 | 444 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |