Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107223.4 | XP_001107223.2 | 509 | 179 | ||
XM_028852050.1 | XP_028707883.1 | 493 | 179 | |||
GenBank mRNA | JU346180.1 | - | 2489 | 179 | ||
JV719510.1 | - | 2485 | 179 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |