Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086222.4 | XP_001086222.2 | 821 | 65 | ||
GenBank mRNA | JV647653.1 | - | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |