Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110332.2 | XP_014965818.2 | 1465 | 186 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |