Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112428.3 | XP_001112428.1 | 1183 | 84 | ||
XM_028827763.1 | XP_028683596.1 | 1450 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |