Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEDD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257441.1 | NP_001244370.1 | 558 | 74 | ||
XM_015113058.2 | XP_014968544.1 | 615 | 74 | |||
XM_015113066.2 | XP_014968552.1 | 625 | 74 | |||
XM_015113067.2 | XP_014968553.1 | 666 | 74 | |||
XM_015113074.2 | XP_014968560.1 | 1982 | 74 | |||
XM_015113077.2 | XP_014968563.1 | 644 | 74 | |||
XM_015113091.2 | XP_014968577.1 | 661 | 74 | |||
GenBank mRNA | JU337080.1 | - | 558 | 74 | ||
JV668551.1 | - | 639 | 74 |
Gene Symbol: NIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU478233.1 | - | 600 | 74 | ||
JV048911.1 | - | 545 | 74 |