Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266272.2 | NP_001253201.1 | 667 | 64 | ||
GenBank mRNA | JU319665.1 | - | 662 | 64 | ||
JU473877.1 | - | 798 | 64 | |||
JV045006.1 | - | 671 | 64 | |||
JV600550.1 | - | 847 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |