Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057098.1 | NP_476439.1 | 2 - 3 | 209 | 76 | |
XM_006235740.1 | XP_006235802.1 | 2 - 3 | 286 | 76 | ||
XM_006235741.1 | XP_006235803.1 | 2 - 3 | 286 | 76 | ||
XM_006235742.2 | XP_006235804.1 | 2 - 3 | 260 | 76 | ||
GenBank mRNA | D12927.1 | - | 2 - 3 | 209 | 76 | |
FQ213752.1 | - | 2 - 3 | 187 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |