Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022239.2 | NP_071575.1 | 2 - 3 | 294 | 108 | |
XM_006250872.2 | XP_006250934.1 | 2 - 3 | 1114 | 108 | ||
GenBank mRNA | M94555.1 | - | 2 - 3 | 294 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |