Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256097.2 | XP_006256159.1 | 3 - 4 | 1684 | 166 |
Gene Symbol: LOC100909544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772773.1 | XP_008770995.1 | 3 - 4 | 1682 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |