Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009605.1 | NP_001009605.1 | 57 | 80 | ||
GenBank mRNA | BC088469.1 | - | 57 | 7128111 | 80 |
Gene Symbol: LOC100911841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749038.3 | XP_003749086.1 | 1 - 2 | 117 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |