Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015004.1 | NP_001015004.1 | 1 - 2 | 433 | 102 | |
GenBank mRNA | BC091399.1 | - | 1 - 2 | 433 | 7323935 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |