Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109499.2 | NP_001102969.1 | 1 - 2 | 127 | 111 | |
XM_006249190.2 | XP_006249252.1 | 2 - 3 | 213 | 111 | ||
XM_006249191.2 | XP_006249253.1 | 2 - 3 | 213 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |