Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012050.1 | NP_001012050.1 | 4 - 5 | 807 | 185 | |
XM_006253091.2 | XP_006253153.1 | 4 - 5 | 610 | 185 | ||
GenBank mRNA | BC088452.1 | - | 4 - 5 | 807 | 7126921 | 185 |
FQ232328.1 | - | 4 - 5 | 864 | 185 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |