Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021849.1 | NP_068621.1 | 3 - 4 | 423 | 152 | |
XM_006247910.2 | XP_006247972.1 | 3 - 4 | 489 | 152 | ||
XM_008768480.1 | XP_008766702.1 | 3 - 4 | 489 | 152 | ||
XM_008768481.1 | XP_008766703.1 | 3 - 4 | 489 | 152 | ||
GenBank mRNA | AB016486.1 | - | 3 - 4 | 423 | 152 | |
BC062031.1 | - | 3 - 4 | 468 | 5623611 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |