Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011963.1 | NP_001011963.1 | 4 - 5 | 663 | 98 | |
XM_006233912.2 | XP_006233974.1 | 4 - 5 | 631 | 98 | ||
XM_008761641.1 | XP_008759863.1 | 3 - 4 | 1397 | 98 | ||
GenBank mRNA | BC079099.1 | - | 4 - 5 | 663 | 7124433 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |