Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK398149.1 | - | 11 | 978 | 78 | |
DQ178124.1 | - | 1 | 46 | 78 | ||
DQ845173.1 | - | 7 | 690 | 78 | ||
KJ786424.1 | - | 10 | 907 | 78 | ||
U82261.1 | - | 4 | 388 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Amplicon spans exons and probe does not span exons |