Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244741.1 | NP_001231670.1 | 5 - 6 | 2054 | 77 | |
XM_005663826.3 | XP_005663883.1 | 1493 | 77 | |||
GenBank mRNA | BE031594.1 | - | 1 - 2 | 40 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |