Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203653.1 | NP_988984.1 | 1 - 2 | 159 | 72 | |
GenBank mRNA | BC061335.1 | - | 1 - 2 | 159 | 72 | |
CT030612.1 | - | 1 - 2 | 100 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |